missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IFN-alpha A Recombinant Protein

Mouse IFN-alpha A Recombinant Protein

Marke:  Gibco™ PMC4016

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 10364303

  • 1128.00 € / 1 Stück
Verfügbar ab: 30-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

Mouse IFN-alpha A Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IFN-α is involved in anti-viral and anti-parasitic activities.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by titration with the use of the cytopathic effect inhibition assay

Shipping Condition: Dry Ice.

Spezifikation

Spezifikation

Cell Culture
19.3 kDa
IFN-α
Store in ultra-cold freezer (-68 to -85°C).
Dry Ice
Cytokines & Receptors
Liquid
Immunology, Virology, Oncology, Signal Transduction
95 %
111654
1 x 10 exp. 5 units
Mouse
10^5 units
E. coli
Recombinant, Ligand
Interferons
Gibco™
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.