Learn More
Gibco™ Rat IL-10 Recombinant Protein
Rat IL-10 Recombinant Protein
Marke: Gibco™ PRC0104
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Rat IL-10 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL10 inhibits the synthesis of a number of cytokines, such as TNF-α, IL-1, and IL-6 from macrophages and IL-2, IFN-γ, and TNF-β from Th1 cells.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Spezifikation
Cell Culture | |
18.7 kDa | |
10 ug | |
Rat | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Inflammation, Signal Transduction | |
98 % |
25325 | |
Affinity Chromatography | |
IL-10 | |
Store in freezer (-5 to -30°C). | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.