missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse CCL22 (MDC) Recombinant Protein
Mouse CCL22 (MDC) Recombinant Protein
Marke: Gibco™ PMC1575
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Mouse CCL22 (MDC) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
MDC displays chemotactic activity for monocytes, dendritic cells, natural killer cells and for chronically activated T lymphocytes. It also displays a mild activity for primary activated T lymphocytes and has no chemoattractant activity for neutrophils, eosinophils and resting T lymphocytes. This chemokine may play a role in the trafficking of activated T lymphocytes to inflammatory sites and other aspects of activated T lymphocyte physiology.Shipping Condition: Wet Ice.
Spezifikation
Cell Culture | |
7.8 kDa | |
20 ug | |
Mouse | |
< 0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology | |
98 % |
11488 | |
Sequential Chromatography | |
MDC | |
Store in freezer (-5 to -30°C). | |
20 μg | |
E. coli | |
Recombinant, Ligand | |
Other Proteins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.