Learn More
Gibco™ Human PDGF-AB Recombinant Protein
Human PDGF-AB Recombinant Protein
Marke: Gibco™ PHG0134
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
Human PDGF-AB Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Members of the PDGF family are mitogenic factors for cells of mesenchymal origin.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Spezifikation
Cell Culture | |
25.5 kDa | |
10 ug | |
Human | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Stem Cell Research, Angiogenesis, Oncology | |
98 % |
5154, 5155 | |
Sequential Chromatography | |
PDGF-AB | |
Store in freezer (-5 to -30°C). | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
PDGFs (Platelet-Derived Growth Factors) | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.