Learn More
Gibco™ Human CCL25 (TECK) Recombinant Protein
Human CCL25 (TECK) Recombinant Protein
Marke: Gibco™ PHC1625
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Human CCL25 (TECK) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
TECK displays chemotactic activity for dendritic cells, thymocytes, and activated macrophages but is inactive on peripheral blood lymphocytes and neutrophils.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Spezifikation
Cell Culture | |
14.2 kDa | |
20 ug | |
127 aa | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Chemokines & Receptors | |
Lyophilized | |
Neurobiology, Immunology | |
98 % |
6370 | |
Sequential Chromatography | |
TECK | |
Human | |
Carrier-Free | |
20 μg | |
E. coli | |
Recombinant, Ligand | |
Other Proteins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.