Learn More
GID4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560684
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DTGNSYLCGY LKIKGLTEEY PTLTTFFEGE IISKKHPFLT RKWDADEDVD RKHWGKFLAF YQYAKSFNSD DFDYE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
GID4 Polyclonal antibody specifically detects GID4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
GID4 | |
Polyclonal | |
Unconjugated | |
GID4 | |
4933439F18Rik; C17orf39; C19H17orf39; C5H17orf39; GID complex subunit 4; GID complex subunit 4 homolog; GID complex subunit 4 homolog; glucose-induced degradation protein 4 homolog; GID complex subunit 4, VID24 homolog; GID complex subunit 4, VID24 homolog (S. cerevisiae); GID4; Glucose-induced degradation protein 4 homolog; im:7147459; LOC509503 protein; LOW QUALITY PROTEIN: glucose-induced degradation protein 4 homolog; vacuolar import and degradation 24; vacuolar import and degradation protein 24 homolog; VID24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79018 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVV7 | |
GID4 | |
Recombinant protein corresponding to Human GID4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only