missing translation for 'onlineSavingsMsg'
Learn More

GIMA7 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA554170

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15782101

  • 530.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: MAESEDRSLR IVLVGKTGSG KSATANTILG EEIFDSRIAA QAVTKNCQKA SREWQGRDLL VVDT Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.

GIMA7 Polyclonal antibody specifically detects GIMA7 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

GIMA7
Polyclonal
Unconjugated
GIMAP7
GIMAP7; GTPase IMAP family member 7; GTPase, IMAP family member 7; hIAN7; IAN7; IAN-7; immune associated nucleotide; immune-associated nucleotide-binding protein 7; immunity-associated nucleotide 7 protein
Rabbit
Antigen affinity chromatography
RUO
168537
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NHV1
GIMAP7
Recombinant protein corresponding to Human GIMA7
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only