Learn More
GIN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557938
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NGDLHLKQIA YYKRTGEYHS TTLPSERSGI RRAAKKFVFK EKKLFYVGKD RKQNRLVIVS EEEKKKVLRE CHENDSGAHH GISRTLT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
GIN1 Polyclonal antibody specifically detects GIN1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
GIN1 | |
Polyclonal | |
Unconjugated | |
GIN1 | |
4930429M06Rik; 8430411H10Rik; Gin1; Gin-1; gypsy retrotransposon integrase 1; gypsy retrotransposon integrase-like protein 1; TGIN1; Ty3/Gypsy integrase 1; ZH2C2; Zinc finger H2C2 domain-containing protein; zinc finger, H2C2 domain containing | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54826 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXP7 | |
GIN1 | |
Recombinant protein corresponding to Human GIN1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only