Learn More
GIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554470
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KKEGHFSALP SLPVGSHAKV SSPQPRGPRY AEGTFISDYS IAMDKIHQQD FVNWLLAQKG KKND Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 69%.
GIP Polyclonal antibody specifically detects GIP in Human samples. It is validated for Immunohistochemistry.Spezifikation
GIP | |
Polyclonal | |
Unconjugated | |
Gip | |
gastric inhibitory polypeptide; Gip; Glucose-dependent insulinotropic peptide; glucose-dependent insulinotropic polypeptide; Gludins; incretin hormone; Rabbit Anti GIP; RATGLUDINS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2695 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09681 | |
Gip | |
Recombinant protein corresponding to Human GIP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only