missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GluR2/GluR3/GluR4 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 11576131
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11576131 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11576131 Lieferant Invitrogen™ Lieferanten-Nr. PA517314

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to a family of glutamate receptors that are sensitive to alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate (AMPA), and function as ligand-activated cation channels. These channels are assembled from 4 related subunits, Gria1-4. The subunit encoded by this gene (Gria2) is subject to RNA editing (Q/R and R/G), which is thought to render the channels impermeable to Ca(2+), and to affect the kinetic aspects of these channels in rat brain. Alternative splicing, resulting in transcript variants encoding different isoforms (flip and flop), has been noted for this gene.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen GluR2/GluR3/GluR4
Anwendungen Western Blot
Klassifikation Polyclonal
Konzentration 79 μg/mL
Konjugat Unconjugated
Zusammensetzung 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gen GRIA3
Gen-Zugriffsnummer P19491, P19492, P19493, P23819, P42262, P42263, P48058, Q9Z2W8, Q9Z2W9
Gen-Alias 2900064I19Rik; AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; AMPA-selective glutamate receptor 3; AMPA-selective glutamate receptor 4; dJ1171F9.1; GluA2; GluA3; GluA4; Glur2; Glur-2; GLUR3; gluR-3; GLUR4; Glur-4; GLUR4C; Gluralpha3; Gluralpha4; GLURB; gluR-B; GLURC; GluR-C; GLURD; gluR-D; GluR-K2; GluR-K3; glutamate ionotropic receptor AMPA type subunit 2; glutamate ionotropic receptor AMPA type subunit 3; glutamate ionotropic receptor AMPA type subunit 4; glutamate receptor; glutamate receptor 2; Glutamate receptor 3; glutamate receptor 4; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor C; glutamate receptor channel alpha 4; glutamate receptor channel alpha3 subunit; glutamate receptor ionotropic, AMPA 2; Glutamate receptor ionotropic, AMPA 3; Glutamate receptor ionotropic, AMPA 4; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotrophic, AMPA 4; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA 4; glutamate receptor, ionotropic, AMPA2 (alpha 2); glutamate receptor, ionotropic, AMPA3 (alpha 3); glutamate receptor, ionotropic, AMPA4 (alpha 4); Gria2; Gria3; Gria4; HBGR2; Kiaa4184; MRX94; spike wave 1; spkw1
Gensymbole GRIA2, GRIA3, GRIA4
Wirtsspezies Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Ser720 of human GluR2.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 14800, 14802, 2891, 2892, 2893, 29627, 29628, 29629, 53623
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.