missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLUT4 Monoclonal Antibody (1F8)
GREENER_CHOICE

Artikelnummer. 11584613
Change view
Click to view available options
Menge:
50 μg
Packungsgröße:
50 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11584613 50 μg 50 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11584613 Lieferant Invitrogen™ Lieferanten-Nr. MA183191

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

This antibody does not react with dog. This product requires enzyme-mediated antigen retrieval prior to staining of paraffin sections. Proteinase K is recommended. Mouse anti glucose transporter 4 antibody, clone 1F8 originally raised against rat intracellular low density microsomes (James et al.

This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen GLUT4
Anwendungen Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon 1F8
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.09% sodium azide; pH 7.2
Gen Slc2a4
Gen-Zugriffsnummer P14142, P14672, P19357
Gen-Alias glucose transporter 4; Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; glucose transporter GLUT4; glucose transporter type 4; glucose transporter type 4, insulin-responsive; GLUT4; GLUT-4; GT2; insulin responsive glucose transporter; insulin-responsive glucose transporter; insulin-responsive glucose transporter type 4; MGC93607; SLC2A4; solute carrier family 2 member 4; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; solute carrier family 2 member 4; solute carrier family 2, facilitated glucose transporter member 4
Gensymbole Slc2a4
Wirtsspezies Mouse
Immunogen Partially purified vesicles containing insulin-responsive glucose transporter 4.
Reinigungsverfahren Protein G
Menge 50 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 100037710, 20528, 25139, 396754, 6517
Zielspezies Human, Mouse, Rat, Rabbit, Monkey, Pig
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.