Learn More
Glyceryltriheptadecanoat, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Glyceryl Triheptadecanoate |
| CAS | 2438-40-6 |
| Summenformel | C54H104O6 |
| Menge | 250 mg |
| Synonym | Heptadecanoic Acid 1,2,3-Propanetriyl Ester, Tri-heptadecanoin, Intarvin, Glycerol Triheptadecanoate, Glyceryl Margarate, Heptadecanoic Acid Triglyceride, Palmitic Acid Triglyderide, Triglyceride HdHdHd, Triheptadecanoin, Trimargarin, Trimargarine, Trimargaroylglycerol |
| InChI-Formel | InChI=1S/C54H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-52(55)58-49-51(60-54(57)48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)50-59-53(56)47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h51H,4-50H2,1-3H3 |
| SMILES | O=C(CCCCCCCCCCCCCCCC)OC(COC(CCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O |
| IUPAC-Name | 2,3-Di(heptadecanoyloxy)propylheptadecanoat |
| Molekulargewicht (g/mol) | 849.4 |
| Formelmasse | 848.7833 |