missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin A / CD235a Mouse, Clone: JC159, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 2993-MSM7-P1
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Glycophorin A / CD235a | |
Cell surface | |
JC159 | |
Unconjugated | |
NP_002091 | |
Mouse | |
39kDa | |
Protein A/G Purified | |
RUO | |
2994 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
GYPA | |
Formalin fixed membrane from a Hairy cell leukemia | |
Erythrocytes in any human tissues | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |