missing translation for 'onlineSavingsMsg'
Learn More

GMPPA Antibody, DyLight 755, Novus Biologicals™

Product Code. 18091157 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18091157 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18091157 Supplier Novus Biologicals Supplier No. NBP297140IR

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GMPPA Polyclonal specifically detects GMPPA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMPPA
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat DyLight 755
Verdünnung Immunohistochemistry, Immunohistochemistry-Paraffin
Zusammensetzung 50mM Sodium Borate
Gen-Alias EC 2.7.7, EC 2.7.7.13, GDP-mannose pyrophosphorylase Amannose-1-phosphate guanyltransferase alpha, GTP-mannose-1-phosphate guanylyltransferase alpha, mannose-1-phosphate guanylyltransferase (GDP)
Wirtsspezies Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GMPPA fragment.
Reinigungsverfahren Antigen Affinity-purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Lipid and Metabolism
Primär oder sekundär Primary
Gen-ID (Entrez) 29926
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.