missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Ig Mix Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen H17000
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Human Ig Mix Polyclonal antibody specifically detects Human Ig Mix in Human samples. It is validated for Flow Cytometry.Spezifikation
Human Ig Mix, Goat anti-Human Ig (H+L) Secondary Antibody | |
Polyclonal | |
PBS with 0.1% sodium azide | |
purified | |
ASR | |
Human | |
Liquid |
Flow Cytometry | |
Unconjugated | |
Goat | |
2 mL | |
Secondary | |
4° C, store in dark | |
IgG |