Learn More
Goat anti-Human IgM Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 31778
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Product # 31778 has been successfully used in Western blot, IF, ICC, IHC and ISH applications. Product # 31778 reacts with the heavy chains of human IgM, but does not react against non-immunoglobulin serum proteins. Based on solid-phase enzyme immunoassay, this antibody cross-reacts with rabbit IgM, swine IgM (25%) rat IgM (12%) guinea pig IgG (6%) mouse IgM (3%) dog IgM (1%), and it has minimal cross-reactivity with immunoglobulins from other species.
Human IgM Polyclonal antibody specifically detects Human IgM in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.Spezifikation
Human IgM | |
Polyclonal | |
Biotin | |
Goat | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
IgG |
Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot | |
0.5 mg/mL | |
PBS with 1mg/mL BSA and 0.08% sodium azide; pH 7.8 | |
purified | |
RUO | |
Human | |
Liquid |