missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG (H+L) Secondary Antibody, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen PA128755
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-28755 detects Mouse IgG from mouse samples.
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Mouse IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
IgG fraction | |
RUO | |
Mouse | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
0.05M tris HCl with 0.0001MM ZnCl2, 0.001M MgCl2, 0.15M NaCl, 10mg/mL BSA, 50% glycerol and 0.1% sodium azide; pH 8 | |
Full length native mouse IgG (purified). | |
100 μg | |
Secondary | |
4° C, do not freeze | |
IgG |