missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 405, Invitrogen™

Goat Polyclonal Secondary Antibody

Marke:  Invitrogen 35500BID

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11895115

  • 176.00 € / 500 Mikroliter
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Product # 35500 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35500 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by solid-phase adsorbed to ensure minimal cross-reactivity with human, bovine, horse, rabbit, swine, goat and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 405 Amax= 400 nm; Emax= 420 nm. Mole Dye/Mole Protein Ratio is lot-dependent.

Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

Mouse IgG (H+L) Cross-Adsorbed
Polyclonal
DyLight 405
Goat
500 μL
Secondary
4° C
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.02% sodium azide; pH 7.2
Antigen affinity chromatography
RUO
Mouse
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote