Learn More
Goat anti-Mouse IgG2a Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A10685
Weitere Details : Netto-Gewicht : 0.10500kg
Beschreibung
Storage and reconstitution: when stored desiccated at or below -20°C, the lyophilized powder is stable for at least one year. Reconstitute using 200 μL PBS, pH 7.2, to yield a 1 mg/mL stock solution containing 0.02% thimerosal. After reconstitution, store product at 4°C. For prolonged storage after reconstitution, add glycerol to a final concentration of 50% (v/v), aliquot, and store at or below -20°C. When stored properly, the solution is stable for approximately three months.
Mouse IgG2a Polyclonal antibody specifically detects Mouse IgG2a in Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Mouse IgG2a | |
Polyclonal | |
PBS with 0.02% thimerosal | |
Mouse IgG2a | |
200 μg | |
Secondary | |
-20°C | |
IgG |
ELISA, Immunohistochemistry, Western Blot | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Lyophilized |