Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 31346
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Product # 31346 has been successfully used in Western blot, IHC and IP applications. Product # 31346 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 0.2 mL of distilled water (1 mg/mL after restoration).
Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Rabbit IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Lyophilized |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
TBS with 15mg/mL BSA, 50mM sucrose and no preservative; pH 8.0 | |
Purified Rabbit IgG, whole molecule | |
0.2 mL | |
Secondary | |
4° C | |
IgG |