Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke:  Invitrogen 31820
Weitere Details : Netto-Gewicht : 1.81440kg
Artikelnummer. 11543350
  • 209.00 € / 1.50 Milligramm
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Product # 31820 has been successfully used in Western blot, IF, ICC, ISH, and IHC applications. Product # 31820 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins. This antibody cross-reacts with rabbit IgM (25%). However, this antibody may cross-react with immunoglobulins from other species.
Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.
Spezifikation
Spezifikation
Rabbit IgG (H+L)
Polyclonal
Biotin
Goat
1.5 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
IgG
Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot
1.5 mg/mL
PBS with 3mg/mL BSA and 0.08% sodium azide; pH 7.8
Affinity chromatography