Learn More
Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 633, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 35563
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Product # 35563 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35563 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by solid-phase adsorbed to ensure minimal cross-reactivity with human, horse, mouse, swine, goat, bovine, and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 633 Amax= 638 nm; Emax= 658 nm. Mole Dye/Mole Protein Ratio is lot-dependent.
Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Rabbit IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 633 | |
Goat | |
500 μL | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
Antigen affinity chromatography | |
RUO | |
Rabbit | |
Liquid |