missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (H+L) Secondary Antibody, DyLight™ 350, Invitrogen™

Goat Polyclonal Secondary Antibody

Marke:  Invitrogen 62270

141.34 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : CAS : 9048-46-8 Netto-Gewicht : 0.01000kg

Artikelnummer. 11889933

  • 212.00 € / 1 Milliliter
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Product # 62270 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 62270 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 350 Amax= 353 nm; Emax= 432 nm. Mole Dye/Mole Protein Ratio is lot-dependent.

Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

Rabbit IgG (H+L)
Polyclonal
DyLight 350
Goat
Antigen affinity chromatography
RUO
Rabbit
Liquid
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 1% BSA and 0.02% sodium azide; pH 7.2
Purified Rabbit IgG, whole molecule
1 mL
Secondary
4° C
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote