missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG F(ab')2 Secondary Antibody, FITC, Invitrogen™

Goat Polyclonal Secondary Antibody

Marke:  Invitrogen 31573

143.34 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.80000kg

Artikelnummer. 11809180

  • 215.00 € / 2 Milligramm
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31573 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31573 reacts with the light chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against the Fc portion of rabbit IgG or against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 7.7 μg/mg; ∽3 moles FITC per mole IgG. (lot-dependent). Reconstitute with 1.5 mL of distilled water. Country of Origin: USA

Rabbit IgG F(ab')2 Polyclonal antibody specifically detects Rabbit IgG F(ab')2 in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.
Spezifikation

Spezifikation

Rabbit IgG F(ab')2
Polyclonal
FITC
Goat
2 mg
Secondary
4° C, store in dark
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
1.5 mg/mL
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Affinity chromatography
RUO
Rabbit
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote