missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™

Goat Polyclonal Secondary Antibody

Marke:  Invitrogen 31583

Weitere Details : Netto-Gewicht : 0.80000kg

Artikelnummer. 11839180

  • 209.00 € / 1.50 Milligramm
Verfügbar ab: 30-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31583 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31583 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 8.8 μg/mL; > 3 moles FITC per mole IgG (lot-dependent). Reconstitute with 1.1 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA

Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.
Spezifikation

Spezifikation

Rabbit IgG (H+L) Cross-Adsorbed
Polyclonal
FITC
Goat
1.5 mg
Secondary
4° C, store in dark
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
1.5 mg/mL
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Antigen affinity chromatography
RUO
Rabbit
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote