Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 31635
Weitere Details : CAS : 17740-40-8 Netto-Gewicht : 2.72160kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31635 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31635 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorescein Amax= 492 nm; Emax= 520 nm. Fluorophore/Protein: 9.3 μg/mg; > 3 moles FITC per mole IgG (lot-dependent). Reconstitute with 1.5 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA
Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.Spezifikation
Rabbit IgG (H+L) | |
Polyclonal | |
FITC | |
Goat | |
2 mg | |
Secondary | |
4° C, store in dark | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
1.5 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Affinity chromatography | |
RUO | |
Rabbit | |
Lyophilized |