Learn More
Invitrogen™ Goat anti-Rat IgG F(ab')2 Secondary Antibody, HRP
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot, and ICC applications. Antibody Specificity: This antibody reacts with the light chains on rat IgG and with those common to other rat immunoglobulins. It does not react with the Fc portion of rat IgG. No antibody was detected against non-immunoglobulin serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 2.0mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Spezifikation
Spezifikation
| Antigen | Rat IgG F(ab')2 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 0.8 mg/mL |
| Konjugat | HRP |
| Zusammensetzung | PBS with 15mg/mL BSA and no preservative; pH 7.6 |
| Wirtsspezies | Goat |
| Reinigungsverfahren | Antigen affinity chromatography |
| Menge | 2 mL |
| Regulatorischer Status | RUO |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.