Learn More
Goat anti-Rat IgM Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 31832
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31832 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31832 reacts with the heavy chain of rat IgM, but not with IgG or with non-immunoglobulin serum proteins. However, antibodies may cross-react with IgM from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 2.0 mL of distilled water. Country of Origin: USA
Rat IgM Polyclonal antibody specifically detects Rat IgM in Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Rat IgM | |
Polyclonal | |
Biotin | |
Goat | |
2 mL | |
Secondary | |
4° C | |
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.8 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Affinity chromatography | |
RUO | |
Rat | |
Lyophilized |