missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat antiserum to Bovine Serum (Forensic) Polyclonal Secondary Antibody, MP Biomedicals
Marke: MP Biomedicals 0855998
Weitere Details : Netto-Gewicht : 0.00200kg
Beschreibung
Host Species: Goat
Species Reactivity: Bovine
Spezifikation
Whole Serum | |
Unconjugated | |
2 mL | |
Bovine |
Polyclonal | |
Goat | |
Secondary | |
Serum |