missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Golgicide A
Potent, selective and reversible inhibitor of GBF1 ArfGEF
N.V.
Spezifikation
Reinheit | >98% |
---|---|
Produkttyp | Golgicide A |
Molekulargewicht | 284.3 |
Hemmstoffe | ARF GTPase Inhibitors |
Beschreibung
Potent, specific and reversible inhibitor of Golgi BFA resistance factor 1 (GBF1), an ArfGEF, that decreases Arf1 activation in vivo. Arrests secretion of soluble and membrane-associated proteins at the endoplasmic reticulum-Golgi intermediate compartment. Causes disassembly and dispersal of the Golgi and trans-Golgi network.Spezifikation
>98% | |
Golgicide A | |
284.3 | |
ARF GTPase Inhibitors |
For Research Use Only