missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GOT2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16396694
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16396694 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16396694 Lieferant Invitrogen™ Lieferanten-Nr. PA577990

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, A375, NIH-3T3, JC, BCL-1, C2C12, Raw264.7 Predicted Reactivity: Rat (97%), Xenopus laevis (82%), Dog (94%), Pig (95%), Chicken (84%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GOT2 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen GOT2
Anwendungen Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 1.33 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen GOT2
Gen-Zugriffsnummer P00505, P00507, P05202
Gen-Alias AL022787; aspartate aminotransferase 2; aspartate aminotransferase precursor (EC 2.6.1.1); aspartate aminotransferase, mitochondrial; aspartate transaminase 2; ASPATA; FABP-1; FABPpm; FABP-pm; fatty acid-binding protein; FLJ40994; Glutamate oxaloacetate transaminase 2; glutamate oxaloacetate transaminase 2, mitochondrial; Glutamate oxaloacetate transaminase 2, mitochondrial (aspartate aminotransferase 2); glutamatic-oxaloacetic transaminase 2, mitochondrial; glutamic-oxaloacetic transaminase 2; glutamic-oxaloacetic transaminase 2, mitochondrial; glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase 2); GOT2; Got-2; KAT4; KATIV; KYAT4; kynurenine aminotransferase 4; Kynurenine aminotransferase IV; kynurenine--oxoglutarate transaminase 4; Kynurenine--oxoglutarate transaminase IV; Maat; mAspAT; mitAAT; mitochondrial aspartate aminotransferase; plasma membrane fatty acid binding protein; plasma membrane-associated fatty acid-binding protein; Transaminase A
Gensymbole GOT2
Wirtsspezies Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GOT2. The exact sequence is proprietary.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 14719, 25721, 2806
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.