Learn More
GPR128 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16863275 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16863275 20 μL 20µL
16853275 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16863275 Supplier Proteintech Supplier No. 247841AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.
Specifications
Antigen GPR128
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ADGRG7
Gene Accession No. Q96K78
Gene Alias G protein coupled receptor 128, GPR128
Gene Symbols ADGRG7
Host Species Rabbit
Immunogen GPR128 Fusion Protein Ag20477
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84873
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG