missing translation for 'onlineSavingsMsg'
Learn More

GRK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 24606-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16883095

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 03-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The sensation of sight is the result of a cascade of events starting with the interaction of photoactivated rhodopsin with a protein called transducin. Rhodopsin kinase is a G-protein-coupled Ser/Thr kinase, also known as GRK1, which is a key element in the regulation of this cascade. Following phosphorylation by GRK1, arrestin is recruited to phospho-rhodopsin quenching its phototransductive activity by preventing further interaction with transducin. By breaking the cycle of phototransduction, GRK1 plays an important role in the restoration of the system for subsequent visual events.
Spezifikation

Spezifikation

GRK1
Polyclonal
Unconjugated
GRK1
GPRK1, GRK1, Rhodopsin kinase, RHOK, RK
Rabbit
Antigen Affinity Chromatography
RUO
24013, 6011
-20°C
Liquid
Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15835, Q9WVL4
GRK1
GRK1 Fusion Protein Ag18063
20 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote