Learn More
GRP94 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL488-14700-100UL
Weitere Details : Netto-Gewicht : 0.00020kg
Beschreibung
The GRP94 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GRP94. This antibody recognizes human, mouse, rat antigen. The GRP94 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Spezifikation
GRP94 | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Flow Cytometry (Intracellular Staining) | |
CoraLite 488 | |
100 μL | |
7184 | |
IgG |