missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSK-3B (GSK3 beta) Recombinant Human Protein
GSK3B is a proline-directed serine/threonine protein kinase.
Marke: Invitrogen™ PR5074A
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
GSK3B is involved in energy metabolism, neuronal cell development and body pattern formation.
Spezifikation
NP_002084 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
GSK-3B (GSK3 beta) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
53.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
GSK Kinase Family | |
Recombinant | |
Baculovirus |