missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GTF2H4 (Human) Recombinant Protein
Human GTF2H4 full-length ORF ( AAH04935.1, 1 a.a. - 462 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00002968-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MESTPSRGLNRVHLQCRNLQEFLGGLSPGVLDRLYGHPATCLAVFRELPSLAKNWVMRMLFLEQPLPQAAVALWVKKEFSKAQEESTGLLSGLRIWHTQLLPGGLQGLILNPIFRQNLRIALLGGGKAWSDDTSQLGPDKHARDVPSLDKYAEERWEVVLHFMVGSPSAAVSQDLAQLLSQAGLMKSTEPGEPPCITSAGFQFLLLDTPAQLWYFMLQYLQTAQSRGMDLVEILSFLFQLSFSTLGKDYSVEGMSDSLLNFLQHLREFGLVFQRKRKSRRYYPTRLAINLSSGVSGAGGTVHQPGFIVVETNYRLYAYTESELQIALIALFSEMLYRFPNMVVAQVTRESVQQAIASGITAQQIIHFLRTRAHPVMLKQTPVLPPTITDQIRLWELERDRLRFTEGVLYNQFLSQVDFELLLAHARELGVLVFENSAKRLMVVTPAGHSDVKRFWKRQKHSS
Spezifikation
AAH04935.1 | |
general transcription factor IIH, polypeptide 4, 52kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GTF2H4 | |
GST |
2968 | |
Wheat germ expression system | |
25 μg | |
TFB2, TFIIH | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |