missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-2K[k] Mouse anti-Mouse, FITC, Clone: 36-7-5, BD
Mouse Monoclonal Antibody
665.00 €
Spécification
Antigen | H-2K[k] |
---|---|
Klon | 36-7-5 |
Konzentration | 0.5mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Code produit | Marque | Menge | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Menge | Prix | Quantité et disponibilité | |||||
15827298
|
BD Biosciences
553592 |
0.5 mg |
665.00 €
1 Stück |
Verfügbar ab: 16-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Description
The 36-7-5 antibody reacts with the H-2K[k] MHC class I alloantigen. Cross-reactivity with splenocytes of SJL/Hsd mice has not been observed by flow cytometric analysis. Reactivity with other haplotypes (e.g., b, d, q) has not been observed.
Spécification
H-2K[k] | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
A.AL mouse splenocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
36-7-5 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
For Research Use Only.