missing translation for 'onlineSavingsMsg'
Learn More

H 89 dihydrochloride, Tocris Bioscience™

Protein kinase A inhibitor

Marke:  Tocris (Bio-Techne) 291050

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412034

  • 934.35 € / 50 Milligramm


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Protein kinase A inhibitor that also inhibits several other kinases (IC 50 values are 80, 120, 135, 270, 2600 and 2800 nM for S6K1, MSK1, PKA, ROCKII, PKBα and MAPKAP-K1b). Exhibits antinociceptive activity. Also available as part of the PKA Tocriset TM .
Spezifikation

Chemische Identifikatoren

130964-39-5
537.294
h-89 dihydrochloride hydrate, n-2-p-bromocinnamylamino ethyl-5-isoquinolinesulfonamide dihydrochloride, c20h20brn3o2s.2clh.h2o, h-89 dihydrochloride hydrate hplc , powder, n-2-2e-3-4-bromophenyl prop-2-en-1-yl amino ethyl isoquinoline-5-sulfonamide hydrate dihydrochloride
N-[2-[[(E)-3-(4-bromophenyl)prop-2-enyl]amino]ethyl]isoquinoline-5-sulfonamide;hydrate;dihydrochloride
C20H24BrCl2N3O3S
GKFFJFGBWAGAFD-HZBIHQSRSA-N
16078954
C1=CC2=C(C=CN=C2)C(=C1)S(=O)(=O)NCCNCC=CC3=CC=C(C=C3)Br.O.Cl.Cl

Spezifikation

H 89 dihydrochloride
130964-39-5
h-89 dihydrochloride hydrate, n-2-p-bromocinnamylamino ethyl-5-isoquinolinesulfonamide dihydrochloride, c20h20brn3o2s.2clh.h2o, h-89 dihydrochloride hydrate hplc , powder, n-2-2e-3-4-bromophenyl prop-2-en-1-yl amino ethyl isoquinoline-5-sulfonamide hydrate dihydrochloride
C1=CC2=C(C=CN=C2)C(=C1)S(=O)(=O)NCCNCC=CC3=CC=C(C=C3)Br.O.Cl.Cl
537.294
519.28
50mg
C20H24BrCl2N3O3S
GKFFJFGBWAGAFD-HZBIHQSRSA-N
N-[2-[[(E)-3-(4-bromophenyl)prop-2-enyl]amino]ethyl]isoquinoline-5-sulfonamide;hydrate;dihydrochloride
16078954
>99%
Videos