missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HAI-2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | HAI-2/SPINT2 |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Anwendungen | Western Blot, Immunoprecipitation |
Klassifikation | Polyclonal |
Beschreibung
HAI-2/SPINT2 Polyclonal specifically detects HAI-2/SPINT2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Spezifikation
HAI-2/SPINT2 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2 | |
SPINT2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
O43291 | |
10653 | |
Mouse myeloma cell line NS0-derived recombinant human HAI-2A Ala28-Lys197 Accession # O43291 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human HAI-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human HAI-1 and recombinant mouse HAI-2B is observed. |
For Research Use Only