missing translation for 'onlineSavingsMsg'
Learn More

Beiersdorf™ Hansaplast Spray Plaster

Product Code. 10323512
Encompass_Preferred
Change view
Click to view available options
Quantity:
32.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
10323512 32.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10323512 Supplier Beiersdorf™ Supplier No. 1861
 Restricted Item
Verfügbar ab: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Hansaplast Spray Plaster
Quantity 32.5 mL
For Use With (Application) Ideal for sports and leisure
compliance-icons compliance-icons
Product Identifier
  • Hansaplast / Elastoplast Sprühpflaster
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable gas.
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P251-Do not pierce or burn, even after use.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
Supplemental information
  • MIXTURE LIST-Contain: ethyl acetate, pentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.