missing translation for 'onlineSavingsMsg'
Learn More

HAPLN2 Rabbit anti-Human, Porcine, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 27818-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16885845

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 03-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

BRAL1 is a member a superfamily consisting of several highly homologous hyaluro and proteoglycan binding link proteins. BRAL1 is predomitly expressed in brain tissue and spinal cord. Like other members in the link-module superfamily, BRAL1 contains an immunoglobulin-like fold and two proteoglycan tandem repeats (PTRs). Its mRNA expression pattern is similar to other lectican proteoglycans, suggesting that BRAL1 may act to stabilize the binding between the extracellular matrix molecule hyaluro and brevican. Immunostaining of mouse brain showed BRAL1 expression at P20 in the white matter of the developing cerebellum and in myelinated fiber tracts in the adult brain, suggesting that expression starts when axonal myelination occurs.
Spezifikation

Spezifikation

HAPLN2
Polyclonal
Unconjugated
HAPLN2
Brain link protein 1, BRAL1, HAPLN2
Rabbit
Antigen Affinity Chromatography
RUO
100628124, 60484, 64057
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9ESM2, Q9GZV7
HAPLN2
HAPLN2 Fusion Protein Ag27169
20 μL
Primary
Porcine, Human, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote