missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16375-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis.Spezifikation
HARS | |
Polyclonal | |
Unconjugated | |
HARS | |
HARS, HisRS, Histidine tRNA ligase, histidyl tRNA synthetase, HRS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15115, 3035, 307492 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12081, Q61035 | |
Hars | |
HARS Fusion Protein Ag9333 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |