missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16824-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Hemoglobin subunit delta is a protein that in humans is encoded by the HBD gene. The delta (HBD) and beta (HBB) genes are normally expressed in the adult: two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin. Two alpha chains plus two delta chains constitute HbA-2, which with HbF comprises the remaining 3% of adult hemoglobin. Five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon-Ggamma-Agamma-delta-beta-3′.Spezifikation
HBD | |
Polyclonal | |
Unconjugated | |
HBD | |
Delta globin, HBD, Hemoglobin delta chain, Hemoglobin subunit delta, hemoglobin, delta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100187828 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02042 | |
HBD | |
HBD Fusion Protein Ag10409 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |