missing translation for 'onlineSavingsMsg'
Learn More

HBZ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 17284-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887154

  • 112.71 € / 20 Mikroliter

Voraussichtliches Versanddatum: 25-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Zeta-globin is an alpha-like hemoglobin. The zeta-globin polypeptide is synthesized in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal and adult like. The zeta-globin gene is a member of the human alpha-globin gene cluster that includes five functional genes and two pseudogenes. The order of genes is: 5′ - zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 -alpha-1 - theta1 - 3′. Zeta-globin (HBZ ) is an alpha-like hemoglobin. The zeta-globin polypeptide is synthesized in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal and adult like. The zeta-globin gene is a member of the human alpha-globin gene cluster that involves 4 functional genes and 3 nonfunctional pseudogenes. The order of genes is: 5′-zeta -- pseudozeta -- pseudoalpha2 -- pseudoalpha1 -- alpha2 -- alph1 -- theta1-3′.
Spezifikation

Spezifikation

HBZ
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02008, P06467
HBAZ, HBZ, HBZ2, Hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, zeta, Zeta globin
Rabbit
IgG
20 μL
-20°C
Primary
15126, 287168, 3050
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HBZ
Liquid
Hbz
HBZ Fusion Protein Ag11061
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate