missing translation for 'onlineSavingsMsg'
Learn More

HDGF Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

N.V.

Spezifikation

Antigen HDGF
Immunogen E. coli-derived recombinant human HDGF Ser2-Lys240 (Pro101Lys) Accession # P51858
Gen-ID (Entrez) 3068
Gen-Zugriffsnummer P51858
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

HDGF Polyclonal specifically detects HDGF in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

HDGF
3068
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects human HDGF in direct ELISAs and Western blots.
Unconjugated
Western Blot
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
E. coli-derived recombinant human HDGF Ser2-Lys240 (Pro101Lys) Accession # P51858
P51858
Reconstitute at 0.2 mg/mL in sterile PBS.
Goat
IgG
Affinity Purified
RUO
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only