Learn More
Brannan Heavy Duty Anemometer with Temperature and Humidity Meter
Beschreibung
- Measures air velocity and air volume with thermometer and humidity meter
- Large LCD with green, yellow and red light indication
- Supplied with calibration certificate and sturdy protective pouch
- Air velocity level indication
- Max min / Avg / data hold function
- Low battery indication
- Audible alarm
- Auto power off
Spezifikation
Spezifikation
| Typ | Anemometer/Thermometer |
| Genauigkeit | ±3% (Full Scale) |
| Luftstromrate | 0.4 to 35 m/sec. (1.4 to 122.5 km/hr.) |
| Gehäusematerial | ABS Plastic |
| Float Material | Polypropylene |
| Gastyp | Oxygen |
| Nenndruck | 100 psig |
| Display | LCD |
| Zur Verwendung mit (Anwendung) | Wind Speed Meter |
| Kennwerte der Stromversorgung | 3 x AA Batteries |