Learn More
Heparin Sodium Salt, TRC
Product Code. 30132863
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132863 2.5 g 2.5g
17949849 1 g 1g
17939749 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132863 Supplier Toronto Research Chemicals Supplier No. TRCH2458002.5G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9041-08-1
Synonym Adomiparin Sodium, Alfa 87-120, Alfa 87-163, Alfa 87-198, Alfa 87-81, Alfa 88-247, Ardeparin Sodium, Badyket, Bemiparin Sodium, Clivarin 1750, Dalteparin Sodium, Deligoparin Sodium, Depo-Heparin, Fragmin, Fragmin IV, H 2149, Hed-Heparin, Hepaflush, Hepagumin, Hepalean, Heparin Fragment Kabi 2165, Heparin sodium, Heparine Choay, Hepathrom, Heprinar, Hepsal, Inno-Hep, Ivor, Kabi 2165, LHN 1, Lioton 1000, Lipo-Hepin, Lipo-Hepinette, Liquaemin Sodium, Liquemin, Logiparin, Longheparin, M 118 Sodium, Minihep, Minolteparin Sodium, Monoparin, Nadroparine Sodium, Normiflo, OP 2000, Panheprin, Parnaparin Sodium, Pularin, RO 11, RO 14, RP 54563, Reviparin Sodium, Semuloparin Sodium, Sevuparin Sodium, Sodium Acid Heparin, Sodium Heparin, Sodium Heparinate, Sodium Parnaparin, Thrombo-Hepin, Thromboliquine, Tinzaparin Sodium, Unihep, Viatromb, WY 90493RD, Zibo
Specifications
Quantity 2.5 g
Chemical Name or Material Heparin Sodium