Learn More
Heptahelicen, TRC
Specifications
Specifications
| Chemischer Name oder Material | Heptahelicene |
| CAS | 16914-68-4 |
| Summenformel | C30 H18 |
| Menge | 1 mg |
| Synonym | Dinaphtho[2,1-c:1',2'-g]phenanthrene |
| InChI-Formel | InChI=1S/C30H18/c1-3-7-25-19(5-1)9-11-21-13-15-23-17-18-24-16-14-22-12-10-20-6-2-4-8-26(20)28(22)30(24)29(23)27(21)25/h1-18H |
| SMILES | c1ccc2c(c1)ccc3ccc4ccc5ccc6ccc7ccccc7c6c5c4c23 |
| IUPAC-Name | Heptacyclo[16.12.0.02,15.03,12.04,9.021,30.024,29]triaconta-1,3(12),4,6,8,10,13,15,17,19,21(30),22,24,26,28-pentadecaen |
| Molekulargewicht (g/mol) | 378.464 |
| Formelmasse | 378.141 |