Learn More
HHIPL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562678
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GRLMSLQENP GTGQWQYSEI CMGHGQTCEF PGLINNYYPY IISFG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
HHIPL1 Polyclonal antibody specifically detects HHIPL1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
HHIPL1 | |
Polyclonal | |
Unconjugated | |
HHIPL1 | |
1600002O04Rik; AK129450; ARAR9245; hedgehog interacting protein-like 1; HHIP like 1; HHIP2; HHIPL1; HHIP-like 1; HHIP-like protein 1; KIAA1822; LOW QUALITY PROTEIN: HHIP-like protein 1; mKIAA1822; RGD1311698; UNQ9245; UNQ9245/PRO34761 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84439 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JK4 | |
HHIPL1 | |
Recombinant protein corresponding to Human HHIPL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only