Learn More
HHLA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563650
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ISNLKTVDPA KFPTRYCYCL NNRTNDLSDF TALLVDIIGN STSYLTEIFK STSILSVNQS NESDCIFICV MTGKSGRNLS DFWEI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
HHLA1 Polyclonal antibody specifically detects HHLA1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
HHLA1 | |
Polyclonal | |
Unconjugated | |
HHLA1 | |
HERV-H LTR-associating 1; HERV-H LTR-associating protein 1; HHLA1; PLA2L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10086 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
C9JL84 | |
HHLA1 | |
Recombinant protein corresponding to Human HHLA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only